Learn More
Purolite™ PuroSorb™ PAD Macroporous Adsorbents
PuroSorb™ adsorbents are macroporous, rigid, highly cross-linked synthetic polymers. They have a variety of base matrices, porosities (or pore volumes), surface areas, and pore sizes. They also exhibit non-polar or hydrophobic tendencies so they adsorb hydrophobic organic compounds.
Marca: Purolite™ LS07360-94
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
- Stable in acidic or alkaline solutions and in most organic solvents
- Regenerate under mild conditions for prolonged life
- Chemically stable, insoluble
- Spherical beads
- Macroporous structure
- Made of styrene, DVB or methacrylate
- Stevia Purification
- Peptide extraction
- Decolorization
- Macrolide purification
- Compliant with ResAP(2004) 3
- Halal and TSE/BSE/GMO free
- Compliant with FDA regulation 21 CFR 173.65
Specifica
Surface Area: 400 to 510 m2/g | |
PAD610 | |
1 kg | |
350 to 1200 μm | |
Wet | |
50 Å | |
Adsorbent |
White to off-white spherical beads | |
Macroporous Polymethacrylic/DVB Adsorbent | |
1 to 14 pH | |
Use within 5 years from date of manufacture, store at 2°C to 20°C | |
60 to 71% | |
Wet |