Learn More
Cole-Parmer™ PVDF Manifold, 10 port
Expand your flow and line capacity with these manifolds. Choose the number of ports and connections to suit your needs.
Marca:  Cole-Parmer™ 31523-55
Dettagli aggiuntivi : Peso : 0.80000kg
Codice prodotto. 11705298
  • 315.00€ / Pezzo
Spedizione stimata: 21-08-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Constructed of durable PVDF.
  • Ideal for applications requiring excellent chemical compatibly.
  • Available in a variety of configurations
Specifica
Specifica
Manifold
1.75 in.
-65°F
1/4 in. x 1/8 in.
1/8 in. NPT(F)
1 in.
1 in.
230°F
PVDF
19.69 cm
-54°C
1/4 in. NPT(F)
2.54 cm
2.54 cm
110 psi
110°C