Learn More
Sartorius Qualitative Grade 1291 Filter Papers
These qualitative filter papers are essentially used for analytical purposes and routine analyses, whenever no gravimetric analyses are required. They are wet-strengthened and can be used for pressure and vacuum filtration.
Marca: Sartorius FT-3-209-185
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.Specifica
Disc | |
180 sec. | |
100/Pk. | |
185 mm |
2 to 3 μm | |
1291 | |
0.16 mm |