Learn More
R&D Systems™ MagCellect Human CD14+ Cell Isolation Kit
Designed to isolate CD14+ cells via a positive selection principle
Marca:  R&D Systems™ MAGH105
Dettagli aggiuntivi : Peso : 0.09700kg
Codice prodotto. 18411480
  • 873.63€ / 1 set
Spedizione stimata: 03-06-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
CD14 is a 55kDa GPI-linked cell surface glycoprotein that is preferentially expressed on monocytes/macrophages. The amino acid sequence of mature human CD14 is approximately 67% and 64% identical to the mouse and rat proteins, respectively.
Typical recovery ranges between 45% and 75% and the purity of recovered CD14+ cells ranges between 90 and 97%
Monocytes isolated using the MagCellect CD14+ Cell Isolation Kit can be used for various downstream applications including the ex vivo generation of dendritic cells
Specifica
Specifica
Store the unopened product at 2°C to 8°C. Do not use past expiration date.