Learn More
R&D Systems™ MagCellect Human NK Cell Isolation Kit
Designed to isolate Natural Killer cells via a negative selection principle
Marca:  R&D Systems™ MAGH109
Dettagli aggiuntivi : Peso : 0.09700kg
Codice prodotto. 18472280
  • 1373.61€ / 1 set
Spedizione stimata: 03-06-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
Typical purity of recovered NK cells ranges between 80% and 90%
MagCellect Streptavidin Ferrofluid is added to the reaction which allows the streptavidin-coated nanoparticles to interact with biotinylated antibody tagged cells, the unwanted cell fraction
Specifica
Specifica
Store the unopened product at 2°C to 8°C. Do not use past expiration date.