Learn More
R&D Systems™ MagCellect Mouse NK Cell Isolation Kit
Designed to isolate Natural Killer cells via a negative selection principle
Marca:  R&D Systems™ MAGM210
Dettagli aggiuntivi : Peso : 0.09700kg
Codice prodotto. 18401360
  • 742.96€ / 1 set
Spedizione stimata: 06-06-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
The purity of recovered cells ranges between 70% and 80%
Magnetically tagged cells will migrate toward the magnet (unwanted cell fraction), leaving the untagged cells or desired cell population in suspension to be harvested by aspiration while the tube remains in the magnetic field
Specifica
Specifica
Store the unopened product at 2°C to 8°C. Do not use past expiration date.