Learn More
RAB6B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10340-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The RAB6B prodtein seems to have a role in retrograde membrane traffic at the level of the Golgi complex. It may also function in retrograde transport in neuronal cells.Specifica
| RAB6B | |
| Polyclonal | |
| Unconjugated | |
| RAB6B | |
| RAB6B, Ras related protein Rab 6B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 270192, 363123, 51560 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P61294, Q9NRW1 | |
| Rab6b | |
| RAB6B Fusion Protein Ag0322 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |