Learn More
Rabbit anti-Chicken IgY (H+L) Secondary Antibody, AP, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marca: Invitrogen SA19514
228.14 EUR valido fino al 2024-05-31
Usa il codice promo "21664" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
This antibody reacts with the heavy and light chains of the immunoglobulin molecule. SA1-9514 has minimum cross-reactivity with IgG of mammalian species. SA1-9514 immunogen is immunoglobulin isolated from chicken serum. Store at 4oC as an undiluted liquid for several weeks. After dilution, do not use for more than one day. For long-term storage, use -20oC.
Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifica
Chicken IgY (H+L) | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
Rabbit | |
Affinity chromatography | |
RUO | |
Chicken | |
Liquid |
ELISA, Western Blot | |
0.667 mg/mL | |
TBS with 10mg/mL BSA, 25% glycerol and 0.02% sodium azide; pH 8.0 | |
Immunoglobulin isolated from chicken serum. | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |