Learn More
Rabbit anti-Goat IgG (H+L) Secondary Antibody, FITC, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marca: Invitrogen SA510313
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
This antibody will react with goat IgG and with light chains common with other goat immunoglobulins as determined by ELISA and IEP techniques. This antibody may cross-react with IgG from other species.
Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifica
Goat IgG (H+L) | |
Polyclonal | |
FITC | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Goat | |
Liquid |
ELISA, Immunocytochemistry, Immunoelectrophoresis, Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Goat IgG h+l | |
1 mg | |
Secondary | |
4° C, store in dark | |
IgG |