Learn More
ACROBiosystems Rabbit CD3E&CD3D Heterodimer Protein, Llama Fc&Llama Fc, low endotoxin (MALS verified)
Marca: ACROBiosystems CDD-R5259-1mg
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
T-cell surface glycoprotein CD3 delta & CD3 epsilon chain, also known as CD3D & CD3E or CD3D&CD3E respectively, are single-pass type I membrane proteins. CD3D, together with CD3- epsilon(CD3E) , CD3-gamma and CD3-zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T cell receptor-CD3 complex. T cell receptor-CD3 complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways.
Rabbit CD3E&CD3D Heterodimer Protein, Llama Fc&Llama Fc is produced by co-expression of CD3E and CD3D, has a calculated MW of 42.4 kDa (CD3E) and 40.8 kDa (CD3D). Subunit CD3E is fused with a llama IgG2b Fc tag at the C-terminus and subunit CD3D is fusedSpecifica
42.4 kDa (CD3E) & 40.8 kDa (CD3D) | |
1 mg | |
Gln 22 - Thr 120 (CD3E) & Thr 22 - Ser 107 (CD3D) | |
CD3E & CD3D,CD3 delta & CD3 epsilon,CD3 delta / epsilon | |
Unconjugated | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers & CD Antigens |
>95% as determined by SDS-PAGE. | |
Rabbit CD3E&CD3D; Heterodimer Protein, Llama Fc&Llama; Fc (CDD-R5259) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Thr 120 (CD3E) & Thr 22 - Ser 107 (CD3D) (Accession # Q9TUF9-1 (CD3E) & A0A5F9D3I4-1 (CD3D)). | |
-20°C | |
CD3E & CD3D | |
Llama IgG Fc Tag | |
Less than 0.01 EU / µg by the LAL method. | |
Powder | |
Rabbit |