missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RABL2A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17816-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This gene is a member of the RAB gene family which belongs to the RAS GTPase superfamily. The proteins in the family of RAS-related signaling molecules are small GTP-binding proteins that play important roles in the regulation of exocytotic and endocytotic pathways. This gene maps to the site of an ancestral telomere fusion event and may be a subtelomeric gene. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifica
| RABL2A | |
| Polyclonal | |
| Unconjugated | |
| RABL2A | |
| Rab like protein 2A, RABL2A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11159, 362987, 68708 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| E9Q9D5, Q9UBK7 | |
| Rabl2, Rabl2A | |
| RABL2A Fusion Protein Ag12135 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto