missing translation for 'onlineSavingsMsg'
Learn More

RAI14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 17507-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16897404

  • 121.38€ / 20µL

Spedizione stimata: 29-09-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

RAI14 plays a role in actin regulation at the ectoplasmic specialization, a type of cell junction specific to testis. Important for establishment of sperm polarity and normal spermatid adhesion. May also promote integrity of Sertoli cell tight junctions at the blood-testis barrier.
Specifica

Specifica

RAI14
Polyclonal
Unconjugated
RAI14
Ankycorbin, KIAA1334, NORPEG, RAI13, RAI14, retinoic acid induced 14
Rabbit
Antigen Affinity Chromatography
RUO
26064, 294804, 75646
-20°C
Liquid
Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5U312, Q9EP71, Q9P0K7
Rai14
RAI14 Fusion Protein Ag11708
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali