missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10469-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Ran (ras-related nuclear protein) is a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The Ran protein is also involved in control of DNA synthesis and cell cycle progression. Nuclear localization of Ran requires the presence of regulator of chromosome condensation 1 (RCC1).Specifica
RAN | |
Polyclonal | |
Unconjugated | |
RAN | |
ARA24, Gsp1, GTPase Ran, RAN, Ras like protein TC4, Ras related nuclear protein, TC4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19384, 5901, 84509 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P62826, P62827, P62828 | |
RAN | |
RAN Fusion Protein Ag0748 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |