missing translation for 'onlineSavingsMsg'
Learn More

RARRES3 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 12065-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16899553

  • 121.38€ / 20µL

Spedizione stimata: 03-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Retinoids exert biologic effects such as potent growth inhibitory and cell differentiation activities and are used in the treatment of hyperproliferative dermatological diseases. These effects are mediated by specific nuclear receptor proteins that are members of the steroid and thyroid hormone receptor superfamily of transcriptional regulators. RARRES1, RARRES2, and RARRES3 are genes whose expression is upregulated by the synthetic retinoid tazarotene. RARRES3 is thought act as a tumor suppressor or growth regulator.
Specifica

Specifica

RARRES3
Polyclonal
Unconjugated
RARRES3
HRASLS4, PLA1/2 3, RAR responsive protein TIG3, RARRES3, RIG1, TIG3
Rabbit
Antigen Affinity Chromatography
RUO
5920
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9UL19
RARRES3
RARRES3 Fusion Protein Ag2702
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali