Learn More
RASEF Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11569-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Binds predomitly GDP, and also GTP.Specifica
RASEF | |
Polyclonal | |
Unconjugated | |
RASEF | |
RAB45, Ras related protein Rab 45, RASEF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
158158, 242505 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5RI75, Q8IZ41 | |
RASEF | |
RASEF Fusion Protein Ag2149 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |