Learn More
RAVER1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA561977
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: LLPEPNILQL LNNLGPSASL QLLLNPLLHG SAGGKQGLLG APPAMPLLNG PALSTALLQL ALQTQGQKKP GILGDSPLG Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
RAVER1 gene ontology annotations related to this gene include RNA binding.Specifica
RAVER1 | |
Polyclonal | |
Unconjugated | |
RAVER1 | |
1300006N24Rik; AA050359; Kiaa1978; LOW QUALITY PROTEIN: ribonucleoprotein PTB-binding 1; mKIAA1978; Protein raver-1; PTB-binding 1; RAVER1; Ribonucleoprotein; Ribonucleoprotein PTB-binding 1; ribonucleoprotein, PTB binding 1; ribonucleoprotein, PTB-binding 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
125950 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.20 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IY67 | |
RAVER1 | |
Recombinant protein corresponding to Human RAVER1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only