Learn More
RBG1L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA563009
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: EVRASLQKVS GSSDSVATMN SEEFVLVPQY ADDNSTKHEE KPQLKIVSNG DEQLE Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 80%.
RABGAP1L is a protein coding gene.Specifica
RBG1L | |
Polyclonal | |
Unconjugated | |
Rabgap1l | |
5830411O09Rik; 8430421H08Rik; 9630005B12Rik; AW049894; expressed in hematopoietic cells, heart, liver (HLL); Hh1; Hhl; Kiaa0471; mKIAA0471; RAB GTPase activating protein 1 like; RAB GTPase activating protein 1-like; rab GTPase-activating protein 1-like; Rab GTPase-activating protein 1-like, isoform 10; RAB GTPase-activating protein 1-like-like protein; rab6 GTPase activating protein-like; RABGAP1L; RGD1304620; TBC1 domain family, member 18; TBC1D18 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9910 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
B7ZAP0 | |
Rabgap1l | |
Recombinant protein corresponding to Human RBG1L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only