Learn More
RCSD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA553455
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: EVPVSFDQPP EGSHLPCYNK VRTRGSIKRR PPSRRFRRSQ SDCGELGDFR AVESSQQNGA KEEDGDEVLP SKSKAPGSPL SSEGAAGEGV RTLGPAEKPP LRRSPSRTEK QEEDRATEEA KNGEK Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 69%.
Stress-induced phosphorylation of CAPZIP may regulate the ability of F-actin-capping protein to remodel actin filament assembly.Specifica
RCSD1 | |
Polyclonal | |
Unconjugated | |
RCSD1 | |
CapZ interacting protein; capZ-interacting protein; CAPZIP; MK2S4; Protein kinase substrate CapZIP; protein kinase substrate MK2S4; RCSD domain containing 1; RCSD domain-containing protein 1; RCSD1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
92241 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6JBY9 | |
RCSD1 | |
Recombinant protein corresponding to Human RCSD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only