Learn More
Honeywell™ RD40 Plastic Filter A2P3
Cost-effective plastic filter protects against hazardous gases, vapors, and particulates.
Marca: Honeywell™ 1788005
Dettagli aggiuntivi : Peso : 0.04500kg
Descrizione
- For use with full-face mask and half-mask equipped with an RD40 connector
- For protection against organic gases and vapors with a boiling point greater than 65°C and solid and liquid particles
- Light and resistant plastic casing
- Suitable for use in metal-free zones and nuclear industry
- Performance requirements for gas and particles significantly above standard for excellent filtration efficiency over extensive period of time
- Canister made of high-grade charcoal, additionally treated for optimal gas absorption
- Specific canister structure for low breathing resistance and increased user comfort
- Low profile for unobstructed field of vision
- Multipurpose: can be used with existing respirators
- Safe storage between periods of use (plastic cover)
Specifica
EC PPE: 3, QA: ISO 9001/2000, EC Cert: 8039 A/15/2 PSA, EC Attestation: 8039 A/15/2 PSA, EN 14387:2004 + A1:2008, Declaration of conformity: APAVE-1788005 | |
285 gr | |
5/Pk. |
RD40 | |
High activated charcoal, ABS plastic with plastic cover | |
106 mm OD |