missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bacteriophage Shiga-like toxin 1 subunit B Protein
A DNA sequence encoding the Enterobacteria phage H19B (Bacteriophage H19B) Shiga-like toxin 1 subunit B, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6941-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Shiga-like toxin 1 subunit B Protein | |
Research Use Only | |
stxB | |
Recombinant Protein | |
E. coli | |
TPDCVTGKVE YTKYNDDDTF TVKVGDKELF TNRWNLQSLL LSAQITGMTV TIKTNACHNG GGFSEVIFR | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Full Length | |
Bacteriophage | |
His-SUMO | |
Tris-based buffer, 50% glycerol |