missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bacteriophage Single-stranded DNA-binding protein gp2.5 Protein
A DNA sequence encoding the Enterobacteria phage T7 (Bacteriophage T7) Single-stranded DNA-binding protein gp2.5, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9695-ye-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_041970.1 | |
Single-stranded DNA-binding protein gp2.5 Protein | |
Research Use Only | |
2.5 | |
Recombinant Protein | |
Yeast | |
MAKKIFTSAL GTAEPYAYIA KPDYGNEERG FGNPRGVYKV DLTIPNKDPR CQRMVDEIVK CHEEAYAAAV EEYEANPPAV ARGKKPLKPY EGDMPFFDNG DGTTTFKFKC YASFQDKKTK ETKHINLVVV DSKGKKMEDV PIIGGGSKLK VKYSLVPYKW NTAVGASVKL QLESVMLVEL ATFGGGEDDW ADEVEENGYV ASGSAKASKP RDEESWDEDD EESEEADEDG DF | |
Greater than 90% as determined by SDS-PAGE. |
1261080 | |
10 μg | |
Not determined. | |
Full Length | |
Bacteriophage | |
His | |
Tris-based buffer, 50% glycerol |