missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine CD8A / MAL Protein
A DNA sequence encoding the Bos taurus (Bovine) CD8A / MAL, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5800-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_776440.1 | |
CD8A / MAL Protein | |
Research Use Only | |
CD8A | |
Recombinant Protein | |
E. coli | |
LSFRMSPTQK ETRLGEKVEL QCELLQSGMA TGCSWLRHIP GDDPRPTFLM YLSAQRVKLA EGLDPRHISG AKVSGTKFQL TLSSFLQEDQ GYYFCSVVSN SILYFSNFVP VFLPAKPATT PAMRPSSAAP TSAPQTRSVS PRSEVCRTSA GSAVDTSRLD FACN | |
Greater than 90% as determined by SDS-PAGE. |
281060 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Bovine | |
His-SUMO | |
Tris-based buffer, 50% glycerol |