missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chicken Riboflavin-binding Protein
A DNA sequence encoding the Gallus gallus (Chicken) Riboflavin-binding protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8851-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_990794.1 | |
Riboflavin-binding Protein | |
Research Use Only | |
RBP | |
Recombinant Protein | |
E. coli | |
QQYGCLEGDT HKANPSPEPN MHECTLYSES SCCYANFTEQ LAHSPIIKVS NSYWNRCGQL SKSCEDFTKK IECFYRCSPH AARWIDPRYT AAIQSVPLCQ SFCDDWYEAC KDDSICAHNW LTDWERDESG ENHCKSKCVP YSEMYANGTD MCQSMWGESF KVSESSCLCL QMNKKDMVAI KHLLSESSEE SSSMSSSEEH ACQKKLLK | |
Greater than 90% as determined by SDS-PAGE. |
396449 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Chicken | |
His | |
Tris-based buffer, 50% glycerol |