missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chimpanzee LTB / TNFSF3 / Lymphotoxin beta Protein
A DNA sequence encoding the Pan troglodytes (Chimpanzee) LTB / TNFSF3 / Lymphotoxin beta, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7788-ec-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001070252.1 | |
LTB / TNFSF3 / Lymphotoxin beta Protein | |
Research Use Only | |
LTB | |
Recombinant Protein | |
E. coli | |
QDQGGLVTET ADPGAQAQQG LGFQKLPEEE PETDLSPGLP AAHLIGAPLK GQGLGWETTK EQAFLTSGTQ FSDAEGLALP QDGLYYLYCL VGYRGRTPPG GGDPQGRSVT LRSSLYRAGG AYGPGTPELL LEGAETVTPV LDPARRQGYG PLWYTSVGFG GLVQLRRGER VYVNISHPDM VDFARGKTFF GAVMVG | |
Greater than 90% as determined by SDS-PAGE. |
744486 | |
100 μg | |
Not determined. | |
Extracellular Domain | |
Chimpanzee | |
His | |
Tris-based buffer, 50% glycerol |