missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chlamydia HSP70 Protein

A cDNA sequence encoding the Chlamydia HSP70 (Amino Acids 549-660) was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP14020-EC-100ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15945429

  • 205.00€ / 100µg

Spedizione stimata: 27-01-2023
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Specifica

Specifica

HSP70 Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Chlamydia
Untagged
Chlamydia HSP70 protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC.
Research Use Only
Recombinant Protein
E. coli
50mM Tris-HCl, pH 8.0, 8M Urea, 60mM NaCl and 50% glycerol.
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali