missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus Dengue-HRP Protein
A cDNA sequence encoding the Dengue-HRP was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11655-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Dengue-HRP Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Phosphate buffered saline, pH 7.4 and 25% glycerol. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Dengue Virus | |
Untagged | |
Greater than 95% as determined by 12% SDS-PAGE (coomassie staining). |