missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog G-CSF / CSF3 Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) G-CSF / CSF3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5878-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
G-CSF / CSF3 Protein | |
Research Use Only | |
CSF3 | |
Recombinant Protein | |
E. coli | |
MAPLGPTGPL PQSFLLKCLE QMRKVQADGT ALQETLCATH QLCHPEELVL LGHALGIPQP PLSSCSSQAL QLMGCLRQLH SGLFLYQGLL QALAGISPEL APTLDTLQLD TTDFAINIWQ QMEDLGMAPA VPPTQGTMPA FTSAFQRRAG GVLVASNLQS FLELAYRALR HFAKP | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Full Length | |
Dog | |
His-SUMO | |
Tris-based buffer, 50% glycerol |