missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog MMP-9 / CLG4B Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) MMP-9 / CLG4B, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9280-ye-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
MMP-9 / CLG4B Protein | |
Research Use Only | |
MMP9 | |
Recombinant Protein | |
Yeast | |
FQTFEGDLKW HHNDITYWIQ NYSEDLPRDV IDDAFARAFA VWSAVTPLTF TRVYGPEADI IIQFGVREHG DGYPFDGKNG LLAHAFPPGP GIQGDAHFDD EELWTLGKGV VVPTHFGNAD GAPCHFPFTF EGRSYSACTT DGRSDDTPWC STTADYDTDR RFGFCPSEKL YAQDGNGDGK PCVFPFTFEG RSYSTCTTDG RSDGYRWCST TADYDQDKLY GFCPTRVDSA VTGGNSAGEP CVFPFIFLGK QYSTCTREGR GDGHLWCATT SNFDRDKKWG FCPDQGYSLF LVAAHEFGHA LGLDHSSVPE ALMYPMYSFT EGPPLHEDDV RGIQHLYGPR PEPEPQPPTA PPTAPPTVCA TGPPTTRPSE RPTAGPTGPP AAGPTGPPTA GPSEAPTVPV DPAEDICKVN IFDAIAEIRN YLHFFKEGKY WRFSKGKGRR VQGPFLSPST WPALPRKLDS AFEDGLTKKT FFFSGRQVWV YTGTSVVGPR RLDKLGLGPE VTQVTGALPQ GGGKVLLFSR QRFWSFDVKT QTVDPRSAGS VEQMYPGVPL NTHDIFQYQE KAYFCQDRFY WRVNSRNEVN QVDEVGYVTF DILQCPED | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Full Length | |
Dog | |
His | |
Tris-based buffer, 50% glycerol |