missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Biosynthetic arginine decarboxylase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Biosynthetic arginine decarboxylase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8804-ec-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_417413.1 | |
Biosynthetic arginine decarboxylase Protein | |
Research Use Only | |
speA | |
Recombinant Protein | |
E. coli | |
DDMSMGLPSS AGEHGVLRSM QEVAMSSQEA SKMLRTYNIA WWGNNYYDVN ELGHISVCPD PDVPEARVDL AQLVKTREAQ GQRLPALFCF PQILQHRLRS INAAFKRARE SYGYNGDYFL VYPIKVNQHR RVIESLIHSG EPLGLEAGSK AELMAVLAHA GMTRSVIVCN GYKDREYIRL ALIGEKMGHK VYLVIEKMSE IAIVLDEAER LNVVPRLGVR ARLASQGSGK WQSSGGEKSK FGLAATQVLQ LVETLREAGR LDSLQLLHFH LGSQMANIRD IATGVRESAR FYVELHKLGV NIQCFDVGGG LGVDYEGTRS QSDCSVNYGL NEYANNIIWA IGDACEENGL PHPTVITESG RAVTAHHTVL VSNIIGVERN EYTVPTAPAE DAPRALQSMW ETWQEMHEPG TRRSLREWLH DSQMDLHDIH IGYSSGIFSL QERAWAEQLY LSMCHEVQKQ LDPQNRAHRP IIDELQERMA DKMYVNFSLF QSMPDAWGID QLFPVLPLEG LDQVPERRAV LLDITCDSDG AIDHYIDGDG IATTMPMPEY DPENPPMLGF FMVGAYQEIL GNMHNLFGDT EAVDVFVFPD GSVEVELSDE GDTVADMLQY VQLDPKTLLT QFRDQVKKTD LDAELQQQFL EEFEAGLYGY TYLE | |
Greater than 90% as determined by SDS-PAGE. |
947432 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |