missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Chaperone SurA Protein
A DNA sequence encoding the Escherichia coli (strain K12) Chaperone SurA, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7328-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_414595.1 | |
Chaperone SurA Protein | |
Research Use Only | |
surA | |
Recombinant Protein | |
E. coli | |
APQVVDKVAA VVNNGVVLES DVDGLMQSVK LNAAQARQQL PDDATLRHQI MERLIMDQII LQMGQKMGVK ISDEQLDQAI ANIAKQNNMT LDQMRSRLAY DGLNYNTYRN QIRKEMIISE VRNNEVRRRI TILPQEVESL AQQVGNQNDA STELNLSHIL IPLPENPTSD QVNEAESQAR AIVDQARNGA DFGKLAIAHS ADQQALNGGQ MGWGRIQELP GIFAQALSTA KKGDIVGPIR SGVGFHILKV NDLRGESKNI SVTEVHARHI LLKPSPIMTD EQARVKLEQI AADIKSGKTT FAAAAKEFSQ DPGSANQGGD LGWATPDIFD PAFRDALTRL NKGQMSAPVH SSFGWHLIEL LDTRNVDKTD AAQKDRAYRM LMNRKFSEEA ASWMQEQRAS AYVKILSN | |
Greater than 90% as determined by SDS-PAGE. |
944812 | |
10 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |