missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Modification methylase EcoRV Protein
A DNA sequence encoding the Escherichia coli (strain K12) Modification methylase EcoRV, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7415-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_863581.1 | |
Modification methylase EcoRV Protein | |
Research Use Only | |
ecoRVM | |
Recombinant Protein | |
E. coli | |
MKDKVFVPPI KSQGIKTKLV PCIKRIVPKN FNGVWVEPFM GTGVVAFNVA PKDALLCDTN PHLISFYNAL KNKDITGDLV KDFLYREGEK LLLSNGEYYY EVRERFNNYK EPLDFLFLNR SCFNGMIRFN SKGGFNVPFC KKPNRFAQAY ITKISNQVDR ISEIISKGNY TFLCQSFEKT IGMVNRDDVV YCDPPYIGRH VDYFNSWGER DERLLFETLS SLNATFITST WHHNDYRENK YVRDLWSSFR ILTKEHFYHV GASEKNRSPM VEALITNIAK DIIDHIEKSS GDILVIEE | |
Greater than 90% as determined by SDS-PAGE. |
1446622 | |
200 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |