missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Ornithine carbamoyltransferase chain F Protein
A DNA sequence encoding the Escherichia coli (strain K12) Ornithine carbamoyltransferase chain F, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8748-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_414807.1 | |
Ornithine carbamoyltransferase chain F Protein | |
Research Use Only | |
argF | |
Recombinant Protein | |
E. coli | |
SDLYKKHFLK LLDFTPAQFT SLLTLAAQLK ADKKNGKEVQ KLTGKNIALI FEKDSTRTRC SFEVAAFDQG ARVTYLGPSG SQIGHKESIK DTARVLGRMY DGIQYRGHGQ EVVETLAQYA GVPVWNGLTN EFHPTQLLAD LMTMQEHLPG KAFNEMTLVY AGDARNNMGN SMLEAAALTG LDLRLLAPKA CWPEESLVAE CSALAEKHGG KITLTEDVAA GVKGADFIYT DVWVSMGEAK EKWAERIALL RGYQVNAQMM ALTDNPNVKF LHCLPAFHDD QTTLGKQMAK EFDLHGGMEV TDEVFESAAS IVFDQAENRM HTIKAVMMAT LGE | |
Greater than 90% as determined by SDS-PAGE. |
944844 | |
50 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |