missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli tsaE Protein
A DNA sequence encoding the Escherichia coli (strain K12) tsaE, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7334-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_418589.1 | |
tsaE Protein | |
Research Use Only | |
tsaE | |
Recombinant Protein | |
E. coli | |
MMNRVIPLPD EQATLDLGER VAKACDGATV IYLYGDLGAG KTTFSRGFLQ ALGHQGNVKS PTYTLVEPYT LDNLMVYHFD LYRLADPEEL EFMGIRDYFA NDAICLVEWP QQGTGVLPDP DVEIHIDYQA QGREARVSAV SSAGELLLAR LAG | |
Greater than 90% as determined by SDS-PAGE. |
948684 | |
10 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |