missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Feline UMOD Protein
A cDNA sequence encoding the UMOD was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13889-2ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
101084248 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Feline | |
MSVCICACGV CFFLCACVCL CTSEVRSCSE CHSNATCMED GVATTCSCQV GFTGDGLTCM DLDECAISGA HNCSEDSICV NMLGSYLCTC PDGFRLMPGL GCTDVDECAE PGLSRCHALA TCVNHKGNYS CVCPAGYWGD GWHCECSPGS CGPGLDCVPE GDTLVCVDPC QAHHILDEYW RSSEYGAGYT CDSGLSGWYR FVGQGGVRLA ETCVPVLSCN TAAPMWLNGT HPSSDEGIVS RTTCAHWRGH CCLWDAPVQV KACAGYYVYN LTAPPECYLA YCTDPSSVLG TCEECSIDED CKMDDGTWSC QCKQDFNITD LSHLERRLEC GANNIKVSLS KCQLKSLGFE KVFMYLRDSQ CSGFIERGDR DWISVVTPAR DGPCGTVMTR NETHATYSNT LYLADEIIIR DLNININFAC SYPLDMKVSL KTSLQPMVST LNISVGGTGM FTVRMALFQS PAYTQPYQGS SVTLATEAFL YVGTMLDGGD LSRFALLMTN CYATPSGNAT DPLRYFIIQD RCPRTKDSTI QVVENGESPQ GRFSVQMFRF AGNYDLVYLH CEVYLCDTVN EKCKPTCSGT RFRSGGIIDQ THVLNLGPIT RKNVQAVVSR AASSSLGFLK VWLPLLLLAS LTLMSQ | |
Greater than 95% as determined by SDS-PAGE. |
UMOD Protein | |
Research Use Only | |
UMOD | |
Feline | |
Untagged | |
The UMOD protein was lyophilized from 0.4microm filtered solution at a concentration of 0.5 mg/ml containing deionized water. |