Learn More
enQuireBio™ Recombinant H. pylori DNA protection during starvation Protein
A DNA sequence encoding the Helicobacter pylori (strain ATCC 700392 / 26695) (Campylobacter pylori) DNA protection during starvation protein, was expressed in the hosts and tags indicated. option, abo
Marca: enQuireBio™ QP7186-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_207041.1 | |
DNA protection during starvation Protein | |
Research Use Only | |
dps | |
Recombinant Protein | |
E. coli | |
MKTFEILKHL QADAIVLFMK VHNFHWNVKG TDFFNVHKAT EEIYEEFADM FDDLAERIVQ LGHHPLVTLS EAIKLTRVKE ETKTSFHSKD IFKEILEDYK YLEKEFKELS NTAEKEGDKV TVTYADDQLA KLQKSIWMLQ AHLA | |
Greater than 90% as determined by SDS-PAGE. |
898765 | |
200 μg | |
Not determined. | |
Full Length | |
H. pylori | |
His-SUMO | |
Tris-based buffer, 50% glycerol |