missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV NS5 Genotype-1a Protein
A cDNA sequence encoding the HCV NS5 Genotype-1a was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12187-GST-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Hepatitis C HCV NS5 Genotype-1a Protein | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
GST |
HCV NS5 Genotype-1a protein is >95% pure as determined by 10% PAGE (coomassie staining). | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Tris, pH 8 and 5mM EDTA. |