missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV NS5 Genotype-2b Protein
A cDNA sequence encoding the HCV NS5 Genotype-2b was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12191-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Hepatitis C HCV NS5 Genotype-2b Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
1.5M urea, 25 mM Tris-HCl, pH 8.0, 0.2% Triton-X and 50% Glycerol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
GST | |
HCV NS5 Genotype-2b protein is >95% pure as determined by 10% PAGE (coomassie staining). |