missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Horse Latherin Protein
A DNA sequence encoding the Equus caballus (Horse) Latherin, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9489-ye-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001075328.2 | |
Latherin Protein | |
Research Use Only | |
LATH | |
Recombinant Protein | |
Yeast | |
QQIPPEVSSQ ITDALTQGLL DGNFLSLLNA INLEGLLNTI LDQVTGLLNI LVGPLLGPSD AEIKLQDTRL LQLSLEFSPD SKGIDIWIPL ELSVYLKLLI LEPLTLYVRT DIRVQLRLES DEDGKYRLAF GHCSLLPRAI ELQSGNPLSL PVNAVLGTIE NALGNFITED LGAGLCPTLN SLVSNLDLQL VNNLINLILD RANVDLSV | |
Greater than 86.7% as determined by SDS-PAGE. |
100033921 | |
10 μg | |
Not determined. | |
Full Length of Mature Protein | |
Horse | |
His | |
Tris-based buffer, 50% glycerol |