missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 3-hydroxyacyl-CoA dehydrogenase type-2 Protein
A DNA sequence encoding the Homo sapiens (Human) 3-hydroxyacyl-CoA dehydrogenase type-2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8037-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001032900.1 | |
3-hydroxyacyl-CoA dehydrogenase type-2 Protein | |
Research Use Only | |
HSD17B10 | |
Recombinant Protein | |
E. coli | |
AAACRSVKGL VAVITGGASG LGLATAERLV GQGASAVLLD LPNSGGEAQA KKLGNNCVFA PADVTSEKDV QTALALAKGK FGRVDVAVNC AGIAVASKTY NLKKGQTHTL EDFQRVLDVN LMGTFNVIRL VAGEMGQNEP DQGGQRGVII NTASVAAFEG QVGQAAYSAS KGGIVGMTLP IARDLAPIGI RVMTIAPGLF GTPLLTSLPE KVCNFLASQV PFPSRLGDPA EYAHLVQAII ENPFLNGEVI RLDGAIRMQP | |
Greater than 90% as determined by SDS-PAGE. |
3028 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |