missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 40S ribosomal protein S24 Protein
A DNA sequence encoding the Homo sapiens (Human) 40S ribosomal protein S24, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8409-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001017.1 | |
40S ribosomal protein S24 Protein | |
Research Use Only | |
RPS24 | |
Recombinant Protein | |
E. coli | |
NDTVTIRTRK FMTNRLLQRK QMVIDVLHPG KATVPKTEIR EKLAKMYKTT PDVIFVFGFR THFGGGKTTG FGMIYDSLDY AKKNEPKHRL ARHGLYEKKK TSRKQRKERK NRMKKVRGTA KANVGAGKKP KE | |
Greater than 90% as determined by SDS-PAGE. |
6229 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |