missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACBD5 Protein
A DNA sequence encoding the Homo sapiens (Human) ACBD5, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7710-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001035938.1 | |
ACBD5 Protein | |
Research Use Only | |
ACBD5 | |
Recombinant Protein | |
E. coli | |
MFQFHAGSWE SWCCCCLIPA DRPWDRGQHW QLEMADTRSV HETRFEAAVK VIQSLPKNGS FQPTNEMMLK FYSFYKQATE GPCKLSRPGF WDPIGRYKWD AWSSLGDMTK EEAMIAYVEE MKKIIETMPM TEKVEELLRV IGPFYEIVED KKSGRSSDIT SVRLEKISKC LEDLGNVLTS TPNAKTVNGK AESSDSGAES EEEEAQEEVK GAEQSDNDKK MMKKSADHKN LEVIVTNGYD KDGFVQDIQN DIHASSSLNG RSTEEVKPID ENLGQTGKSA VCIHQDINDD HVEDVTGIQH LTSDSDSEVY CDSMEQFGQE ESLDSFTSNN GPFQYYLGGH SSQPMENSGF REDIQVPPGN GNIGNMQVVA VEGKGEVKHG GEDGRNNSGA PHREKRGGET DEFSNVRRGR GHRMQHLSEG TKGRQVGSGG DGERWGSDRG SRGSLNEQIA LVLMRLQEDM Q | |
Greater than 90% as determined by SDS-PAGE. |
91452 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |