missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Active regulator of SIRT1 Protein
A DNA sequence encoding the Homo sapiens (Human) Active regulator of SIRT1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8330-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Active regulator of SIRT1 Protein | |
Research Use Only | |
AROS | |
Recombinant Protein | |
E. coli | |
MSAALLRRGL ELLAASEAPR DPPGQAKPRG APVKRPRKTK AIQAQKLRNS AKGKVPKSAL DEYRKRECRD HLRVNLKFLT RTRSTVAESV SQQILRQNRG RKACDRPVAK TKKKKAEGTV FTEEDFQKFQ QEYFGS | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |