missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ADH7 Protein
A DNA sequence encoding the Homo sapiens (Human) ADH7, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5617-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000664.2 | |
ADH7 Protein | |
Research Use Only | |
ADH7 | |
Recombinant Protein | |
E. coli | |
MFAEIQIQDK DRMGTAGKVI KCKAAVLWEQ KQPFSIEEIE VAPPKTKEVR IKILATGICR TDDHVIKGTM VSKFPVIVGH EATGIVESIG EGVTTVKPGD KVIPLFLPQC RECNACRNPD GNLCIRSDIT GRGVLADGTT RFTCKGKPVH HFMNTSTFTE YTVVDESSVA KIDDAAPPEK VCLIGCGFST GYGAAVKTGK VKPGSTCVVF GLGGVGLSVI MGCKSAGASR IIGIDLNKDK FEKAMAVGAT ECISPKDSTK PISEVLSEMT GNNVGYTFEV IGHLETMIDA LASCHMNYGT SVVVGVPPSA KMLTYDPMLL FTGRTWKGCV FGGLKSRDDV PKLVTEFLAK KFDLDQLITH VLPFKKISEG FELLNSGQSI RTVLTF | |
Greater than 90% as determined by SDS-PAGE. |
131 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |