missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ADP-ribosylation factor 6 Protein
A DNA sequence encoding the Homo sapiens (Human) ADP-ribosylation factor 6, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5674-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001654.1 | |
ADP-ribosylation factor 6 Protein | |
Research Use Only | |
ARF6 | |
Recombinant Protein | |
E. coli | |
MGKVLSKIFG NKEMRILMLG LDAAGKTTIL YKLKLGQSVT TIPTVGFNVE TVTYKNVKFN VWDVGGQDKI RPLWRHYYTG TQGLIFVVDC ADRDRIDEAR QELHRIINDR EMRDAIILIF ANKQDLPDAM KPHEIQEKLG LTRIRDRNWY VQPSCATSGD GLYEGLTWLT SNYKS | |
Greater than 90% as determined by SDS-PAGE. |
382 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |