missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Aflatoxin B1 aldehyde reductase member 3 Protein
A DNA sequence encoding the Homo sapiens (Human) Aflatoxin B1 aldehyde reductase member 3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5640-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_036199.2 | |
Aflatoxin B1 aldehyde reductase member 3 Protein | |
Research Use Only | |
AKR7A3 | |
Recombinant Protein | |
E. coli | |
MSRQLSRARP ATVLGAMEMG RRMDAPTSAA VTRAFLERGH TEIDTAFVYS EGQSETILGG LGLRLGGSDC RVKIDTKAIP LFGNSLKPDS LRFQLETSLK RLQCPRVDLF YLHMPDHSTP VEETLRACHQ LHQEGKFVEL GLSNYAAWEV AEICTLCKSN GWILPTVYQG MYNAITRQVE TELFPCLRHF GLRFYAFNPL AGGLLTGKYK YEDKDGKQPV GRFFGNTWAE MYRNRYWKEH HFEGIALVEK ALQAAYGASA PSMTSATLRW MYHHSQLQGA HGDAVILGMS SLEQLEQNLA AAEEGPLEPA VVDAFNQAWH LVAHECPNYF R | |
Greater than 80% as determined by SDS-PAGE. |
22977 | |
500 μg | |
Not determined. | |
Full Length of BC025709 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |