missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AGRP Protein
A DNA sequence encoding the Homo sapiens (Human) AGRP, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5623-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001129.1 | |
AGRP Protein | |
Research Use Only | |
AGRP | |
Recombinant Protein | |
E. coli | |
AQMGLAPMEG IRRPDQALLP ELPGLGLRAP LKKTTAEQAE EDLLQEAQAL AEVLDLQDRE PRSSRRCVRL HESCLGQQVP CCDPCATCYC RFFNAFCYCR KLGTAMNPCS RT | |
Greater than 90% as determined by SDS-PAGE. |
181 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |