missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AIMP1 / EMAP2 / SCYE1 Protein
A DNA sequence encoding the Human AIMP1 / EMAP2 / SCYE1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10366-ec-20ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001135887.1 | |
AIMP1 / EMAP2 / SCYE1 Protein | |
Research Use Only | |
AIMP1 | |
The ED50 as determined by the apoptotic effect using serum free human MCF-7 cells is less than 40 ng/ml, corresponding to a specific activity of > 2.5 M 104 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
9255 | |
20 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SKPIDVSRLD LRIGCIITAR KHPDADSLYV EEVDVGEIAP RTVVSGLVNH VPLEQMQNRM VILLCNLKPA KMRGVLSQAM VMCASSPEKI EILAPPNGSV PGDRITFDAF PGEPDKELNP KKKIWEQIQP DLHTNDECVA TYKGVPFEVK GKGVCRAQTM SNSGIK | |
Greater than 98% as determined by SDS-PAGE and HPLC. |