missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Alpha-tocopherol transfer Protein
A DNA sequence encoding the Homo sapiens (Human) Alpha-tocopherol transfer protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6847-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000361.1 | |
Alpha-tocopherol transfer Protein | |
Research Use Only | |
TTPA | |
Recombinant Protein | |
E. coli | |
MAEARSQPSA GPQLNALPDH SPLLQPGLAA LRRRAREAGV PLAPLPLTDS FLLRFLRARD FDLDLAWRLL KNYYKWRAEC PEISADLHPR SIIGLLKAGY HGVLRSRDPT GSKVLIYRIA HWDPKVFTAY DVFRVSLITS ELIVQEVETQ RNGIKAIFDL EGWQFSHAFQ ITPSVAKKIA AVLTDSFPLK VRGIHLINEP VIFHAVFSMI KPFLTEKIKE RIHMHGNNYK QSLLQHFPDI LPLEYGGEEF SMEDICQEWT NFIMKSEDYL SSISESIQ | |
Greater than 90% as determined by SDS-PAGE. |
7274 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |