missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Annexin A2 Protein
A DNA sequence encoding the Homo sapiens (Human) Annexin A2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5652-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001002857.1 | |
Annexin A2 Protein | |
Research Use Only | |
ANXA2 | |
Recombinant Protein | |
E. coli | |
STVHEILCKL SLEGDHSTPP SAYGSVKAYT NFDAERDALN IETAIKTKGV DEVTIVNILT NRSNAQRQDI AFAYQRRTKK ELASALKSAL SGHLETVILG LLKTPAQYDA SELKASMKGL GTDEDSLIEI ICSRTNQELQ EINRVYKEMY KTDLEKDIIS DTSGDFRKLM VALAKGRRAE DGSVIDYELI DQDARDLYDA GVKRKGTDVP KWISIMTERS VPHLQKVFDR YKSYSPYDML ESIRKEVKGD LENAFLNLVQ CIQNKPLYFA DRLYDSMKGK GTRDKVLIRI MVSRSEVDML KIRSEFKRKY GKSLYYYIQQ DTKGDYQKAL LYLCGGDD | |
Greater than 90% as determined by SDS-PAGE. |
302 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |