missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human APEX1 / AP / APEx / Ref-1 Protein
A DNA sequence encoding the Homo sapiens (Human) APEX1 / AP / APEx / Ref-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP899-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001231178.1 | |
APEX1 / AP / APEx / Ref-1 Protein | |
Research Use Only | |
APEX1 | |
Recombinant Protein | |
E. coli | |
KNDKEAAGEG PALYEDPPDQ KTSPSGKPAT LKICSWNVDG LRAWIKKKGL DWVKEEAPDI LCLQETKCSE NKLPAELQEL PGLSHQYWSA PSDKEGYSGV GLLSRQCPLK VSYGIGDEEH DQEGRVIVAE FDSFVLVTAY VPNAGRGLVR LEYRQRWDEA FRKFLKGLAS RKPLVLCGDL NVAHEEIDLR NPKGNKKNAG FTPQERQGFG ELLQAVPLAD SFRHLYPNTP YAYTFWTYMM NARSKNVGWR LDYFLLSHSL LPALCDSKIR SKALGSDHCP ITLYLAL | |
Greater than 90% as determined by SDS-PAGE. |
328 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |